Gene Transcripts
- Gene Symbol
- ZEB1-AS1
- Entrez Gene ID
- 220930
- Gene Name
- ZEB1 antisense RNA 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 31316528 - 31319095 on Build GRCh38
- UniGene ID
- Hs.372654
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN483252.1 |
- |
2 |
532 |
|
Chr. 10 - 31318025 - 31319032 [-] |
GENCODE |
ENST00000441257.1 |
- |
2 |
517 |
|
Chr. 10 - 31318088 - 31319027 [-] |
GenBankmRNA |
BI518871.1 |
- |
2 |
518 |
|
Chr. 10 - 31318088 - 31319027 [-] |
View Full Product Details