Gene Transcripts
- Gene Symbol
- IDS2
- Entrez Gene ID
- 105373366
- Gene Name
- uncharacterized IDS2
- Gene Aliases
- Chromosome Location
- Chr. X: 149511554 - 149526523 on Build GRCh38
- UniGene ID
- Hs.731041
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF050146.1 |
- |
1 |
121 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000370438.2 |
- |
1 |
121 |
|
Chr. X - 149502660 - 149505280 [-] |
View Full Product Details