Gene Transcripts
- Gene Symbol
- RTN4RL2
- Entrez Gene ID
- 349667
- Gene Name
- reticulon 4 receptor-like 2
- Gene Aliases
- NGRH1, NgR2
- Chromosome Location
- Chr. 11: 57460537 - 57477539 on Build GRCh38
- UniGene ID
- Hs.502618
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
HY323308.1 |
- |
1 |
88 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000528885.1 |
- |
1 |
81 |
|
Chr. 11 - 57476493 - 57477534 [-] |
View Full Product Details