Gene Transcripts
- Gene Symbol
- Eldr
- Entrez Gene ID
- 72735
- Gene Name
- Egfr long non-coding downstream RNA
- Gene Aliases
- 2810442I21Rik, 4930500E12Rik, Fabl
- Chromosome Location
- Chr. 11: 16934709 - 16951282 on Build GRCm38
- UniGene ID
- Mm.268598
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK141825.1 |
- |
1 |
94 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY729982.1 |
- |
1 |
94 |
|
Chr. 11 - 16939187 - 16951093 [-] |
View Full Product Details