Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033541.4 | NP_291019.1 | 2 | 423 | |
GenBank mRNA | AB067528.1 | - | 1 | 80 | |
AF459815.1 | - | 1 | 80 | ||
AF459816.1 | - | 1 | 80 | ||
AK028986.1 | - | 2 | 424 | ||
AK078952.1 | - | 1 | 91 | ||
AK163253.1 | - | 1 | 96 | ||
BC108955.1 | - | 1 | 89 | 40046017 | |
BC126877.1 | - | 1 | 88 | 40046021 |