Gene Transcripts

Gene Symbol
Peg10
Entrez Gene ID
170676
Gene Name
paternally expressed 10
Gene Aliases
AA407948, Edr, HB-1, MEF3L, Mar2, Mart2, MyEF-3
Chromosome Location
Chr. 6: 4747306 - 4760517 on Build GRCm38
UniGene ID
Mm.320575
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040611.1 NP_001035701.1 2 489
NM_130877.2 NP_570947.2 2 489
GenBank mRNA AB091827.1 - 2 415
AJ006464.1 - 2 399

View Full Product Details