Gene Transcripts
- Gene Symbol
- Magea9-ps
- Entrez Gene ID
- 195772
- Gene Name
- melanoma antigen, family A, 9, pseudogene
- Gene Aliases
- Mage-a9, Magea9
- Chromosome Location
- Chr. X: 72869518 - 72870174 on Build GRCm38
- UniGene ID
- Mm.377231
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC116352.1 |
- |
1 |
483 |
|
BC116353.1 |
- |
1 |
485 |
40099901 |
View Full Product Details