Gene Transcripts

Gene Symbol
Kif15
Entrez Gene ID
209737
Gene Name
kinesin family member 15
Gene Aliases
3110023M17Rik, 3930402I10Rik, D330038N01, HKLP2, Knsl7, b2b1117.1Clo, b2b1117.1aClo
Chromosome Location
Chr. 9: 122951052 - 123018733 on Build GRCm38
UniGene ID
Mm.278920
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010620.1 NP_034750.1 21 2659
GenBank mRNA AJ560623.1 - 21 2676
AK052362.1 - 16 2082
BC060710.1 - 21 2659 6409082

View Full Product Details