Gene Transcripts
- Gene Symbol
- Rdh19
- Entrez Gene ID
- 216453
- Gene Name
- retinol dehydrogenase 19
- Gene Aliases
- RDH-S, RDH-S2, Rdhs
- Chromosome Location
- Chr. 10: 127849928 - 127861176 on Build GRCm38
- UniGene ID
- Mm.222953
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_147222.2 |
NP_671755.2
|
3, 4 |
833 |
|
GenBank mRNA |
AY046408.1 |
- |
3, 4 |
829 |
|
View Full Product Details