Gene Transcripts

Gene Symbol
Mlh3
Entrez Gene ID
217716
Gene Name
mutL homolog 3 (E coli)
Gene Aliases
AV125803, BB126472
Chromosome Location
Chr. 12: 85234466 - 85270648 on Build GRCm38
UniGene ID
Mm.311981
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK132195.1 - 2 1385
AK162464.1 - 1 52
BC072599.1 - 2 1395
BC079861.1 - 2 1383 6834767

View Full Product Details