Gene Transcripts

Gene Symbol
Wdr60
Entrez Gene ID
217935
Gene Name
WD repeat domain 60
Gene Aliases
D430033N04Rik
Chromosome Location
Chr. 12: 116206261 - 116263047 on Build GRCm38
UniGene ID
Mm.31382
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146039.3 NP_666151.3 7 1321
GenBank mRNA AK046399.1 - 7 1283
AK052482.1 - 7 1285
AK144138.1 - 7 1321
BC138916.1 - 7 1254

View Full Product Details