Gene Transcripts

Gene Symbol
PHF2
Entrez Gene ID
5253
Gene Name
PHD finger protein 2
Gene Aliases
CENP-35, GRC5, JHDM1E
Chromosome Location
Chr. 9: 93576627 - 93679587 on Build GRCh38
UniGene ID
Hs.211441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005392.3 NP_005383.3 6 795
XM_005252051.2 XP_005252108.1 6 789
XM_006717143.2 XP_006717206.1 6 789
GenBank mRNA AB014562.2 - 5 688
AF043725.1 - 6 1002
AL834263.1 - 6 646
BC152437.1 - 5 688

View Full Product Details