Gene Transcripts

Gene Symbol
Eddm3b
Entrez Gene ID
219026
Gene Name
epididymal protein 3B
Gene Aliases
Fam12, Gm75
Chromosome Location
Chr. 14: 51114370 - 51117815 on Build GRCm38
UniGene ID
Mm.297297
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203508.1 NP_987104.1 2 153
GenBank mRNA AK136724.1 - 2 293
BC031737.1 - 2 153 4221053

View Full Product Details