Gene Transcripts
- Gene Symbol
- Aaas
- Entrez Gene ID
- 223921
- Gene Name
- achalasia, adrenocortical insufficiency, alacrimia
- Gene Aliases
- AAA, ADRACALA, D030041N15Rik, GL003
- Chromosome Location
- Chr. 15: 102338247 - 102350759 on Build GRCm38
- UniGene ID
- Mm.352946
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153416.2 |
NP_700465.2
|
3, 4 |
456 |
|
GenBank mRNA |
AK034591.1 |
- |
3, 4 |
351 |
|
AK079849.1 |
- |
3, 4 |
393 |
|
AK083401.1 |
- |
3, 4 |
369 |
|
AK083537.1 |
- |
3, 4 |
377 |
|
AK087134.1 |
- |
3, 4 |
431 |
|
AK088247.1 |
- |
3, 4 |
373 |
|
AK134406.1 |
- |
3, 4 |
347 |
|
AK165243.1 |
- |
3, 4 |
371 |
|
AK166118.1 |
- |
3, 4 |
371 |
|
AK169523.1 |
- |
3, 4 |
408 |
|
AK189195.1 |
- |
2, 3 |
146 |
|
BC018191.1 |
- |
3, 4 |
357 |
3994248 |
BC025501.1 |
- |
3, 4 |
403 |
5321013 |
BC026604.1 |
- |
2, 3 |
625 |
|
View Full Product Details