Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290495.1 | NP_001277424.1 | 1 | 101 | |
NM_001290496.1 | NP_001277425.1 | 1 | 101 | ||
NM_145520.5 | NP_663495.3 | 1 | 101 | ||
GenBank mRNA | AK043949.1 | - | 1 | 95 | |
AK050583.1 | - | 1 | 83 | ||
AK050587.1 | - | 1 | 91 | ||
AK081579.1 | - | 1 | 91 | ||
AK087102.1 | - | 1 | 83 | ||
AK090004.1 | - | 933 | |||
AK167294.1 | - | 1 | 77 | ||
AU079587.1 | - | 1 | 101 | ||
BC015285.1 | - | 1 | 80 | 4239390 | |
BC024075.1 | - | 1 | 70 | 5097466 | |
BU053791.1 | - | 1 | 53 | ||
BU707805.1 | - | 1 | 63 |