Gene Transcripts

Gene Symbol
Slc25a25
Entrez Gene ID
227731
Gene Name
solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 25
Gene Aliases
1110030N17Rik, MCSC, mKIAA1896
Chromosome Location
Chr. 2: 32414487 - 32451470 on Build GRCm38
UniGene ID
Mm.37395
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164357.1 NP_001157829.1 2, 3 532
NM_001164358.1 NP_001157830.1 2, 3 751
NM_001290558.1 NP_001277487.1 2, 3 401
NM_146118.3 NP_666230.2 2, 3 532
GenBank mRNA AK049392.1 - 2, 3 508
AK082756.1 - 2, 3 489
AK122568.2 - 2, 3 716
AK132204.1 - 2, 3 486
BB653459.1 - 2, 3 533
BC019978.1 - 2, 3 340 4239441
BC022114.1 - 2, 3 338 5098924
BC037109.1 - 2, 3 492 5368306
BC066998.1 - 2, 3 719 5703622

View Full Product Details