Gene Transcripts
- Gene Symbol
- Tardbp
- Entrez Gene ID
- 230908
- Gene Name
- TAR DNA binding protein
- Gene Aliases
- 1190002A23Rik, C85084, TDP-43, Tdp43
- Chromosome Location
- Chr. 4: 148612382 - 148626996 on Build GRCm38
- UniGene ID
- Mm.22453
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003898.3 |
NP_001003898.1
|
4, 5 |
867 |
|
NM_001003899.2 |
NP_001003899.1
|
4, 5 |
867 |
|
NM_001008545.2 |
NP_001008545.1
|
4, 5 |
867 |
|
NM_001008546.2 |
NP_001008546.1
|
4, 5 |
867 |
|
NM_145556.4 |
NP_663531.1
|
4, 5 |
867 |
|
GenBank mRNA |
AK032199.1 |
- |
4, 5 |
637 |
|
AK045495.1 |
- |
4, 5 |
637 |
|
AK083312.1 |
- |
4, 5 |
641 |
|
AK133207.1 |
- |
4, 5 |
644 |
|
AK146002.1 |
- |
4, 5 |
637 |
|
AK146997.1 |
- |
5, 6 |
546 |
|
AK153803.1 |
- |
4, 5 |
641 |
|
AY319513.1 |
- |
3, 4 |
537 |
|
AY319514.1 |
- |
3, 4 |
537 |
|
AY319515.1 |
- |
3, 4 |
537 |
|
AY319516.1 |
- |
3, 4 |
537 |
|
BC012873.1 |
- |
4, 5 |
625 |
4016437 |
BC025544.1 |
- |
4, 5 |
638 |
5321007 |
BC027105.1 |
- |
4, 5 |
629 |
5359082 |
BC027772.1 |
- |
4, 5 |
627 |
5346061 |
BC029728.1 |
- |
4, 5 |
628 |
4950263 |
BC031126.1 |
- |
4, 5 |
629 |
4977114 |
BC033475.1 |
- |
4, 5 |
625 |
4953334 |
GQ428775.1 |
- |
3, 4 |
537 |
|
GQ428776.1 |
- |
3, 4 |
537 |
|
GQ428777.1 |
- |
3, 4 |
537 |
|
View Full Product Details