Gene Transcripts

Gene Symbol
Myo1h
Entrez Gene ID
231646
Gene Name
myosin 1H
Gene Aliases
4631401O15Rik
Chromosome Location
Chr. 5: 114288899 - 114364576 on Build GRCm38
UniGene ID
Mm.238285
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164573.1 NP_001158045.1 17 1689

View Full Product Details