Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145210.2 | NP_660211.2 | 3 | 531 | |
GenBank mRNA | AB067531.1 | - | 3 | 505 | |
AK087823.1 | - | 3 | 531 | ||
AY055830.1 | - | 3 | 504 | ||
AY055831.1 | - | 3 | 504 | ||
BC053721.1 | - | 3 | 528 | 30053184 | |
BC094918.1 | - | 3 | 521 | 30050168 | |
BC108956.1 | - | 3 | 527 | 40046020 |