Gene Transcripts

Gene Symbol
Smyd5
Entrez Gene ID
232187
Gene Name
SET and MYND domain containing 5
Gene Aliases
AW536703, NN8-4AG, Rai15, Rrg1
Chromosome Location
Chr. 6: 85431976 - 85446429 on Build GRCm38
UniGene ID
Mm.219946
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144918.2 NP_659167.2 8 752
GenBank mRNA AK158267.1 - 8 733
AK162926.1 - 8 676
BC016525.1 - 8 739 3601356

View Full Product Details