Gene Transcripts

Gene Symbol
Vmn2r19
Entrez Gene ID
232358
Gene Name
vomeronasal 2, receptor 19
Gene Aliases
EG232358
Chromosome Location
Chr. 6: 123308333 - 123336537 on Build GRCm38
UniGene ID
Mm.469878
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104632.1 NP_001098102.1 3 878

View Full Product Details