Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163756.1 | NP_001157228.1 | 7 | 1088 | |
NM_172759.3 | NP_766347.1 | 7 | 1088 | ||
GenBank mRNA | AK033563.1 | - | 7 | 1051 | |
AK077248.1 | - | 7 | 1079 | ||
BC022148.1 | - | 3 | 401 | ||
BC055062.1 | - | 7 | 1057 | 6528966 |