Gene Transcripts

Gene Symbol
Ces4a
Entrez Gene ID
234677
Gene Name
carboxylesterase 4A
Gene Aliases
Ces8
Chromosome Location
Chr. 8: 105131800 - 105150109 on Build GRCm38
UniGene ID
Mm.120297
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146213.2 NP_666325.2 14 1782
GenBank mRNA AK037191.1 - 14 1705
BC026374.1 - 14 1782 4163362

View Full Product Details