Gene Transcripts

Gene Symbol
Rltpr
Entrez Gene ID
234695
Gene Name
RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing
Gene Aliases
CG1399-PB, D130029J02Rik, Gm585
Chromosome Location
Chr. 8: 105686010 - 105698195 on Build GRCm38
UniGene ID
Mm.471640
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033320.2 NP_001028492.1 11 1558
GenBank mRNA AK031012.1 - 11 1557
BC115830.1 - 3 178
HF678090.1 - 32 3189

View Full Product Details