Gene Transcripts

Gene Symbol
Cpz
Entrez Gene ID
242939
Gene Name
carboxypeptidase Z
Gene Aliases
AV136066
Chromosome Location
Chr. 5: 35502218 - 35525626 on Build GRCm38
UniGene ID
Mm.108557
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153107.2 NP_694747.2 9 1486
GenBank mRNA AF356844.1 - 9 1395
BX518878.1 - 3 313

View Full Product Details