Gene Transcripts

Gene Symbol
Sbno1
Entrez Gene ID
243272
Gene Name
sno, strawberry notch homolog 1 (Drosophila)
Gene Aliases
9330180L10Rik, AI849128, AW557836, BC021875, Sbno, mSno1, sno
Chromosome Location
Chr. 5: 124368702 - 124426011 on Build GRCm38
UniGene ID
Mm.23879
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081203.1 NP_001074672.1 19 2692
GenBank mRNA AB188811.1 - 18 2621
AK133096.1 - 11 1516
BC027253.1 - 1 80
BC138418.1 - 19 2749 8861438

View Full Product Details