Gene Transcripts
- Gene Symbol
- Smok3a
- Entrez Gene ID
- 545814
- Gene Name
- sperm motility kinase 3A
- Gene Aliases
- Smok2b1
- Chromosome Location
- Chr. 5: 138021276 - 138034665 on Build GRCm38
- UniGene ID
- Mm.349005
- Species
- Mus musculus
- Gene Symbol
- Smok3b
- Entrez Gene ID
- 622474
- Gene Name
- sperm motility kinase 3B
- Gene Aliases
- 4931427P12Rik, Smok2b2
- Chromosome Location
- Chr. 5: 138037224 - 138050633 on Build GRCm38
- UniGene ID
- Mm.456691
- Species
- Mus musculus
- Gene Symbol
- Smok4a
- Entrez Gene ID
- 272667
- Gene Name
- sperm motility kinase 4A
- Gene Aliases
- 4930513D10Rik, Smok2c
- Chromosome Location
- Chr. 17: 13521456 - 13528434 on Build GRCm38
- UniGene ID
- Mm.444718
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK029728.1 |
- |
2 |
1393 |
|
BC125398.1 |
- |
1 |
951 |
|
BC132092.1 |
- |
1 |
951 |
|
View Full Product Details