Gene Transcripts

Gene Symbol
Snhg11
Entrez Gene ID
319317
Gene Name
small nucleolar RNA host gene 11
Gene Aliases
A930034L06Rik, AI854265, AL022637, AW319638, E130013N09Rik
Chromosome Location
Chr. 2: 158375638 - 158386145 on Build GRCm38
UniGene ID
Mm.294494
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175692.3 NP_783623.2 2, 3 490
GenBank mRNA AK080745.1 - 2, 3 448
BC030414.1 - 2, 3 406 5366924
BC049241.1 - 404 4505955

View Full Product Details