Gene Transcripts
- Gene Symbol
- Casc1
- Entrez Gene ID
- 320662
- Gene Name
- cancer susceptibility candidate 1
- Gene Aliases
- A230084G12Rik, Las1
- Chromosome Location
- Chr. 6: 145174872 - 145210970 on Build GRCm38
- UniGene ID
- Mm.207779
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_177222.4 |
NP_796196.3
|
4, 5 |
361 |
|
GenBank mRNA |
AK034148.1 |
- |
3, 4 |
209 |
|
AK132912.1 |
- |
4, 5 |
585 |
|
AY423542.1 |
- |
4, 5 |
292 |
|
AY485607.1 |
- |
4, 5 |
361 |
|
AY485608.1 |
- |
4, 5 |
361 |
|
BC072568.1 |
- |
2, 3 |
203 |
|
BC120743.1 |
- |
4, 5 |
265 |
40129666 |
BC125423.1 |
- |
4, 5 |
265 |
40129938 |
BC144801.1 |
- |
4, 5 |
265 |
|
View Full Product Details