Gene Transcripts

Gene Symbol
Pate2
Entrez Gene ID
330921
Gene Name
prostate and testis expressed 2
Gene Aliases
Gm846, Pate-M, mANLP1
Chromosome Location
Chr. 9: 35574309 - 35698689 on Build GRCm38
UniGene ID
Mm.479701
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033421.3 NP_001028593.1 3 192
GenBank mRNA AK136651.1 - 3 193
AK136739.1 - 1 902

View Full Product Details