Gene Transcripts

Gene Symbol
Grxcr2
Entrez Gene ID
332309
Gene Name
glutaredoxin, cysteine rich 2
Gene Aliases
Gm851
Chromosome Location
Chr. 18: 41986201 - 41999049 on Build GRCm38
UniGene ID
Mm.240520
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033426.2 NP_001028598.1 1 104
GenBank mRNA AK158418.1 - 1 105
BC147102.1 - 1 99 9056155
BC147103.1 - 1 99 9056135

View Full Product Details