Gene Transcripts

Gene Symbol
Clec4b2
Entrez Gene ID
381809
Gene Name
C-type lectin domain family 4, member b2
Gene Aliases
Aplra1, F830043G12Rik, mDCAR1
Chromosome Location
Chr. 6: 123173023 - 123204749 on Build GRCm38
UniGene ID
Mm.333372
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004159.2 NP_001004159.1 3, 4 240
GenBank mRNA AK156711.1 - 3, 4 372
AY365133.1 - 3, 4 263

View Full Product Details