Gene Transcripts

Gene Symbol
Zbtb2
Entrez Gene ID
381990
Gene Name
zinc finger and BTB domain containing 2
Gene Aliases
Gm1103
Chromosome Location
Chr. 10: 4367073 - 4392701 on Build GRCm38
UniGene ID
Mm.262466
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033466.3 NP_001028638.2 3 669
GenBank mRNA AK030936.1 - 3 517
AK132308.1 - 3 670
AK160748.1 - 3 499
AK173184.1 - 1 4244
BC094553.1 - 1 97

View Full Product Details