Gene Transcripts

Gene Symbol
Best3
Entrez Gene ID
382427
Gene Name
bestrophin 3
Gene Aliases
Vmd2l3, mBest4
Chromosome Location
Chr. 10: 116986314 - 117025687 on Build GRCm38
UniGene ID
Mm.339322
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007583.1 NP_001007584.1 2, 3 668
GenBank mRNA AY450426.1 - 2, 3 668

View Full Product Details