Gene Transcripts

Gene Symbol
Mrgprb8
Entrez Gene ID
404240
Gene Name
MAS-related GPR, member B8
Gene Aliases
MrgB8
Chromosome Location
Chr. 7: 48388526 - 48389648 on Build GRCm38
UniGene ID
Mm.357942
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207539.2 NP_997422.1 1 569
GenBank mRNA BC138164.1 - 1 569
BC138165.1 - 1 569 8861184

View Full Product Details