Gene Transcripts

Gene Symbol
Gpr31b
Entrez Gene ID
436440
Gene Name
G protein-coupled receptor 31, D17Leh66b region
Gene Aliases
12-HETER, GPR31c(t), Gpr31, Gpr31c
Chromosome Location
Chr. 17: 13051321 - 13052280 on Build GRCm38
UniGene ID
Mm.335670
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013832.2 NP_001013854.2 1 889
GenBank mRNA AK036897.1 - 571

View Full Product Details