Gene Transcripts

Gene Symbol
Zc3h12b
Entrez Gene ID
547176
Gene Name
zinc finger CCCH-type containing 12B
Gene Aliases
A130028J20Rik, CXorf32, Gm373
Chromosome Location
Chr. X: 95711529 - 95932656 on Build GRCm38
UniGene ID
Mm.302708
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034907.2 NP_001030079.2 9 2601

View Full Product Details