Gene Transcripts

Gene Symbol
Mup6
Entrez Gene ID
620807
Gene Name
major urinary protein 6
Gene Aliases
Gm12544, Mup2, OTTMUSG00000007423
Chromosome Location
Chr. 4: 60003549 - 60006214 on Build GRCm38
UniGene ID
Mm.464562
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081285.1 NP_001074754.1 4 328

View Full Product Details