Gene Transcripts
- Gene Symbol
- Trbv1
- Entrez Gene ID
- 621968
- Gene Name
- T cell receptor beta, variable 1
- Gene Aliases
- Gm6273, TCRBV2S1, Tcrb-V2
- Chromosome Location
- Chr. 6: 40891296 - 40891885 on Build GRCm38
- UniGene ID
- Mm.333009
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF479018.1 |
- |
|
182 |
|
AK138971.1 |
- |
|
251 |
|
M16680.1 |
- |
2 |
182 |
|
View Full Product Details