Gene Transcripts

Gene Symbol
Rad54b
Entrez Gene ID
623474
Gene Name
RAD54 homolog B (S. cerevisiae)
Gene Aliases
E130016E03Rik
Chromosome Location
Chr. 4: 11558920 - 11615808 on Build GRCm38
UniGene ID
Mm.491200
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039556.3 NP_001034645.1 4 450
GenBank mRNA BC057604.1 - 4 398

View Full Product Details