Gene Transcripts

Gene Symbol
Vmn1r68
Entrez Gene ID
628580
Gene Name
vomeronasal 1 receptor 68
Gene Aliases
EG628580, Gm6898
Chromosome Location
Chr. 7: 10527210 - 10528169 on Build GRCm38
UniGene ID
Mm.480301
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172072.1 NP_001165543.1 1 553

View Full Product Details