Gene Transcripts

Gene Symbol
Trav9d-4
Entrez Gene ID
667574
Gene Name
T cell receptor alpha variable 9D-4
Gene Aliases
Gm13907
Chromosome Location
Chr. 14: 52983456 - 52983770 on Build GRCm38
UniGene ID
Mm.446617
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC139214.1 - 962
BC139216.1 - 962

View Full Product Details