Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009076.3 | NP_033102.2 | 5 | 408 | |
GenBank mRNA | AK002973.1 | - | 5 | 407 | |
AK008347.1 | - | 5 | 407 | ||
AK012349.1 | - | 5 | 407 | ||
AK012428.1 | - | 5 | 405 | ||
AK088464.1 | - | 5 | 407 | ||
AK144905.1 | - | 5 | 407 | ||
AK148566.1 | - | 1039 | |||
AK150318.1 | - | 5 | 408 | ||
AK151532.1 | - | 5 | 407 | ||
AK151592.1 | - | 5 | 407 | ||
AK167761.1 | - | 5 | 427 | ||
AK168121.1 | - | 5 | 407 | ||
AK198056.1 | - | 3 | 168 | ||
BC018321.1 | - | 5 | 404 | 3583073 | |
BC075731.1 | - | 3 | 1026 | ||
BC081469.1 | - | 5 | 391 | 6824411 | |
BC090393.1 | - | 386 | 5011867 | ||
L04280.1 | - | 5 | 395 |