Gene Transcripts

Gene Symbol
Spint3
Entrez Gene ID
629747
Gene Name
serine peptidase inhibitor, Kunitz type, 3
Gene Aliases
Gm14315, OTTMUSG00000016288
Chromosome Location
Chr. 2: 164569695 - 164573456 on Build GRCm38
UniGene ID
Mm.234248
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177401.1 NP_001170872.1 1 222
GenBank mRNA AK136730.1 - 1 200
BU938823.1 - 1 225

View Full Product Details