Gene Transcripts

Gene Symbol
Skint6
Entrez Gene ID
230622
Gene Name
selection and upkeep of intraepithelial T cells 6
Gene Aliases
OTTMUSG00000008519
Chromosome Location
Chr. 4: 112804503 - 113286973 on Build GRCm38
UniGene ID
Mm.291107
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001103199.1 NP_001096669.1 2 76
GenBank mRNA EU099304.1 - 2 76
EU099305.1 - 2 70

View Full Product Details