Gene Transcripts

Gene Symbol
Pcnxl3
Entrez Gene ID
104401
Gene Name
pecanex-like 3 (Drosophila)
Gene Aliases
mKIAA4073
Chromosome Location
Chr. 19: 5664635 - 5688908 on Build GRCm38
UniGene ID
Mm.329650
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144868.3 NP_659117.2 5 1322
GenBank mRNA AK171516.1 - 4 1561
BC054751.1 - 5 601
BC055289.1 - 6 1125
BC096538.1 - 5 1322

View Full Product Details