Gene Transcripts

Gene Symbol
Ugt1a10
Entrez Gene ID
394430
Gene Name
UDP glycosyltransferase 1 family, polypeptide A10
Gene Aliases
A13, Ugt1a13
Chromosome Location
Chr. 1: 88055411 - 88220002 on Build GRCm38
UniGene ID
Mm.300095
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201641.2 NP_964003.2 1 631
GenBank mRNA AY227201.1 - 1 590
BC089569.1 - 1 631 30120494

View Full Product Details