Gene Transcripts

Gene Symbol
Adrb2
Entrez Gene ID
24176
Gene Name
adrenoceptor beta 2, surface
Gene Aliases
Chromosome Location
Chr. 18: 57513792 - 57515834 on Build Rnor_6.0
UniGene ID
Rn.10206
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012492.2 NP_036624.2 1 776
GenBank mRNA AY057895.1 - 1024
AY057896.1 - 1 1025
BC086538.1 - 1 776 7114502
J03024.1 - 1 678

View Full Product Details