Gene Transcripts
- Gene Symbol
- Cyp2g1
- Entrez Gene ID
- 25251
- Gene Name
- cytochrome P450, family 2, subfamily g, polypeptide 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 83626639 - 83637780 on Build Rnor_6.0
- UniGene ID
- Rn.10909
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012787.1 |
NP_036919.1
|
3 |
400 |
|
GenBank mRNA |
M33296.2 |
- |
3 |
398 |
|
View Full Product Details