Gene Transcripts

Gene Symbol
SLC18A3
Entrez Gene ID
6572
Gene Name
solute carrier family 18 member A3
Gene Aliases
VACHT
Chromosome Location
Chr. 10: 49610301 - 49612720 on Build GRCh38
UniGene ID
Hs.654374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003055.2 NP_003046.2 1 1339
GenBank mRNA AK313094.1 - 1 1228
BC007765.2 - 1 1265 4136084
U09210.1 - 1 1341

View Full Product Details